Magnesium oxide light E530, BP, Ph. Eur., extra pure, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Magnesia usta
1.05862.1000 1.05862.9020
1.05862.1000EA 0 EUR
1.05862.1000 1.05862.9020
Magnesium oxide light E530, BP, Ph. Eur., extra pure, SAFC®
Magnesium oxide
Formule: MgO
MW: 40.3 g/mol
Kookpunt: 3600
Smeltpunt: 2800
Dichtheid: 3.58 g/cm³ (25 °C)
MDL Nummer: MFCD00011109
CAS nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700