Magnesium oxide heavy 98.0-100.5% (by complexometry from ignited basis), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.05867.1000 1.05867.9029
1.05867.1000EA 0 EUR
1.05867.1000 1.05867.9029
Magnesium oxide heavy 98.0-100.5% (by complexometry from ignited basis), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Magnesium oxide
Formule: MgO
Kookpunt: 3600 °C (1013 hPa)
Smeltpunt: 2800 °C
Dichtheid: 3,58 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011109
CAS nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 12,05713