Calcium hydrogen orthophosphate, anhydrous, extra fine powder, EMPROVE® EXPERT FCC, BP, E341, Ph. Eur., USP suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic
1.02144.9026 1.02144.9025 1.02144.1000
1.02144.9026EA 0 EUR
1.02144.9026 1.02144.9025 1.02144.1000
Calcium hydrogen orthophosphate, anhydrous, extra fine powder, EMPROVE® EXPERT FCC, BP, E341, Ph. Eur., USP suitable for use as excipient, SAFC®
Calcium hydrogen orthophosphate
Formule: CaHPO₄
MW: 136.06 g/mol
Smeltpunt: 370
Dichtheid: 2.89 g/cm³ (20 °C)
MDL Nummer: MFCD00010909
CAS nummer: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697