Aluminium oxide substrate 10x10x1 mm, polished one side C plane
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Aluminium(III) oxide, Alumina C, Aluminum oxide
40281.KT 40281.KU
40281.KTEA 157 EUR
40281.KT 40281.KU
Aluminium oxide substrate 10x10x1 mm, polished one side C plane
Aluminium oxide
Molecular Formula: Al2O3 MDL No.: MFCD00003424
Formule: Al₂O₃
MW: 101.96 g/mol
Kookpunt: 2980
Smeltpunt: 2045
Dichtheid: 3.94 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
Bereik: 01-2119529248-35
Merck Index: 13,00355