Aluminium oxide, Durmax® for metallography, 12 hours
Leverancier: ALUMINES DURMAX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20993.266 20993.296
20993.266EA 108 EUR
20993.266 ALUD20993.296
Aluminium oxide, Durmax® for metallography, 12 hours
Aluminium oxide
Leveringsinformatie: This product is not available in all countries. Please check with your local VWR International office or supplier.
Formule: Al₂O₃
Kookpunt: 2980 °C (1013 hPa)
Smeltpunt: 2045 °C
Dichtheid: 3,94 g/cm³ (20 °C)
MDL Nummer: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
Bereik: 01-2119529248-35
Merck Index: 13,00355