Indium(III)oxide, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Indium trioxide, Diindium trioxide
MQ200
1.12201.0010 1.12201.0005
1.12201.0010EA 95.8 EUR
1.12201.0010 1.12201.0005
Indium(III)oxide, Sigma-Aldrich®
Indium(III)oxide
Formule: In₂O₃
MW: 277.64 g/mol
Smeltpunt: 715.05
Dichtheid: 7.18 g/cm³ (20 °C)
MDL Nummer: MFCD00011060
CAS nummer: 1312-43-2
EINECS: 215-193-9
Merck Index: 12,04984
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 2000 °C
Molar Mass 277.64 g/mol
Density 7.18 g/cm3 (20 °C)