Trimethylboraat ≥99.9995% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
22975.09
22975.18
22975.30
22975.09EA
36.4
EUR
22975.09
22975.18
22975.30
Trimethylboraat ≥99.9995% (metals basis)
Trimethylboraat
Purity: 99.9995+%
CAS No.: 121-43-7
Molecular Formula: C3H9BO3
Formula Weight: 103.92
Flash Point: -8°(17°F)
Physical Form: Liquid
Density: 0.915
Refractive Index: 1.3610
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Odourless
Melting Point: -34°
Boiling Point: 67-69°
Beilstein Reference: 1697939
Merck Reference: 14,9712
UN No.: 2416
MDL No.: MFCD00008346
Formule:
C₃H₉BO₃ Kookpunt: 67…69 °C Smeltpunt: –34 °C Dichtheid: 0,915 Vlampunt: –8 °C (17 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008346 CAS nummer: 121-43-7 EINECS: 204-468-9 UN: 2416 ADR: 3,II Merck Index: 14,9712 |