Aluminium ammonium sulphate dodecahydrate ≥99% (by titrimetric analysis), Reagent Grade
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A2140-1KG A2140-500G A2140-10KG
A2140-1KGEA 297 EUR
A2140-1KG A2140-500G HONCA2140-10KG
Aluminium ammonium sulphate dodecahydrate ≥99% (by titrimetric analysis), Reagent Grade
Aluminium ammonium sulphate dodecahydrate
  • ≥95% for general laboratory use.
Formule: AlNH₄(SO₄)₂·12H₂O
Kookpunt: 200 °C (1013 hPa)
Dichtheid: 1.640 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149958
CAS nummer: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325
Order Now
Specification Test Results
Assay Min. 99 %
Solution complying