Lead(II) carbonate ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Lead carbonat
40119.22 40119.36 40119.57
40119.22EA 70.9 EUR
40119.22 40119.36 40119.57
Lead(II) carbonate ACS
Lead(II) carbonate
CAS No.: 598-63-0 Molecular Formula: PbCO3 Formula Weight: 267.20 Physical Form: Powder Odour: Odourless UN No.: 2291 MDL No.: MFCD00064631
Formule: PbCO₃
MW: 267.21 g/mol
Smeltpunt: 315
Dichtheid: 6.43 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064631
CAS nummer: 598-63-0
EINECS: 209-943-4
UN: 2291
ADR: 6.1 III