Microcentrifuge tubes, ClikLok™ and MacrewTube™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T330-76N T330-75N T330-75B T330-75G T330-75R T330-75Y T330-75AM T330-75NS T330-75AMS T330-76B T330-76G T330-76R T330-76Y T330-76AM T330-76NS T330-76AMS T365-5TN T365-5TAM T365-5NLSS
SIMPT330-76NEA 93 EUR
SIMPT330-76N SIMPT330-75N SIMPT330-75B SIMPT330-75G SIMPT330-75R SIMPT330-75Y SIMPT330-75AM SIMPT330-75NS SIMPT330-75AMS SIMPT330-76B SIMPT330-76G SIMPT330-76R SIMPT330-76Y SIMPT330-76AM SIMPT330-76NS SIMPT330-76AMS SIMPT365-5TN SIMPT365-5TAM SIMPT365-5NLSS
Microcentrifuge tubes, ClikLok™ and MacrewTube™
Buisjes Microcentrifugebuisjes
These tubes are made of high-quality, transparent polypropylene without the use of slip agents, plasticisers, mold release agents and biocide substances.
  • RNase-, DNase-, Pyrogen- and DNA-free
  • Writing surface on flat cap and side wall for quick sample identification
  • Precise lid sealing for minimal evaporation rate during long-term storage
  • Graduated every 0,5 ml
  • Exceptional centrifugation stability of up to 25000×g
  • Withstands temperatures from +121 to –196 °C
The ClikLok™ and MacrewTube™ tubes have been designed for the simple and safe processing of medium-sized sample volumes.
Designed for samples larger than 2,0 ml. The 16 mm tube diameter is identical to the diameter of standard 15 ml tubes, ensuring compatibility with centrifuge rotors. Both the ClikLok™ and MacrewTube™ are designed with a smooth bottom with no sharp points to prevent hurting user fingers. The slim design fits high capacity rotors.
The ClikLok™ snap cap tubes are available with a flat top or with a CapInsert™ cavity for color-coding purposes. The cap will partially open when the internal pressure approaches an unsafe level in a controlled manner, releasing excess pressure but preventing any splashing of contents. In applications over 80 °C, the CapLock™ clips are recommended.
The MacrewTube™ screw cap tubes are available sterile or with a choice of two caps, a regular lip seal cap or a tethered cap. Both models have the CapInsert™ cavity for colour coding purposes.
Bestelinformatie: Colour coding CapInserts™, Caplock™ clips, universal adapters, racks and storage boxes are available separately.
Verpakking: Sterile products are packaged in bags of 20. Non-sterile tubes are packaged in bags of 100.