Kieselguhr, EMPROVE® ESSENTIAL NF suitable for use as excipient, purified and calcined, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Diatomaceous silica, Diatomaceous earth, Siliceous earth, Infusorial earth, Diatomite, Celite
1.08117.9025 1.08117.9026
1.08117.9025EA 361 EUR
1.08117.9025 1.08117.9026
Kieselguhr, EMPROVE® ESSENTIAL NF suitable for use as excipient, purified and calcined, SAFC®
Kieselguhr
Formule: O₂Si
Dichtheid: 2,35 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00132803
CAS nummer: 68855-54-9
EINECS: 272-489-0