Tin(II) sulphate, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Tin(II) sulphate, Tin sulphate, Stannous sulphate
M-Clarity™ quality level = MQ200
1.12416.0250 1.12416.1000
1.12416.0250EA 53.8 EUR
1.12416.0250 1.12416.1000
Tin(II) sulphate, Sigma-Aldrich®
Tin(II) sulphate
Formule: O₄SSn
Dichtheid: 4,15 g/cm³ (20 °C)
MDL Nummer: MFCD00011246
CAS nummer: 7488-55-3
EINECS: 231-302-2
Merck Index: 12,08947
Order Now
Specification Test Results
Solubility (20 °C) soluble
Molar Mass 214.77 g/mol
Bulk Density 800 kg/m3
pH 1.6 (50 g/l, H2O, 20 °C)