Dibutyltindichloride for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Dichlorodibutylstannane, Di-n-butyltin dichloride, DBTC
M-Clarity™ quality level = MQ200
8.04184.0250 8.04184.1000 8.04184.9030
8.04184.0250EA 95.4 EUR
8.04184.0250 8.04184.1000 8.04184.9030
Dibutyltindichloride for synthesis, Sigma-Aldrich®
Dibutyltindichloride
Formule: C₈H₁₈Cl₂Sn
Kookpunt: 275 °C (1013 hPa)
Smeltpunt: 37…39 °C
Dichtheid: 1,37…1.4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000518
CAS nummer: 683-18-1
EINECS: 211-670-0
UN: 3146
ADR: 6.1,II
Order Now
Specification Test Results
Solubility 0.32 g/l (20 °C)
Melting Point 37 - 41 °C
Molar Mass 303.85 g/mol
Boiling Point 148 °C (16 hPa)
Vapor Pressure 0.0016 hPa (25 °C)
Flash Point 100 °C
Density 1.37 - 1.40 g/cm3 (20 °C)