Chlorotriethylsilane ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A15547.09
A15547.18
A15547.30
A15547.09EA
42.6
EUR
A15547.09
A15547.18
A15547.30
Chlorotriethylsilane ≥98%
Chlorotriethylsilane
Purity: 98+%
CAS No.: 994-30-9
Molecular Formula: C6H15ClSi
Formula Weight: 150.73
Flash Point: 29°(84°F)
Refractive Index: 1.4310
Odour: Pungent
Melting Point: ca -75°
Boiling Point: 145-147°
Beilstein Reference: 1732860
UN No.: 2986
MDL No.: MFCD00000507
Formule:
C₆H₁₅ClSi Kookpunt: 145…147 °C Smeltpunt: ∼–75 °C Dichtheid: 0,899 Vlampunt: 29 °C (84 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000507 CAS nummer: 994-30-9 EINECS: 213-615-6 UN: 2986 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...