Triethylsilane 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
212920050 212920250 212921000 212925000
ACRO212920050EA 32 EUR
ACRO212920050 ACRO212920250 ACRO212921000 ACRO212925000
Triethylsilane 99%
Triethylsilane
Formule: C₆H₁₆Si
Kookpunt: 107…108 °C (760 mmHg)
Smeltpunt: -156,9 °C
Dichtheid: 0,72 g/cm³
Vlampunt: –3 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009018
CAS nummer: 617-86-7
EINECS: 210-535-3
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20°C, 589 nm)
GC ≥98.5 %