Tetramethyltin 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
163980100 163980500
ACRO163980100EA 71.5 EUR
ACRO163980100 ACRO163980500
Tetramethyltin 98%
Tetramethyltin
Formule: C₄H₁₂Sn
Kookpunt: 75 °C (1013 hPa)
Smeltpunt: –54 °C
Dichtheid: 1,29 g/cm³
Vlampunt: –12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008278
CAS nummer: 594-27-4
EINECS: 209-833-6
UN: 3384
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥97.5 %