Cyclopentyltrimethoxysilane 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L16442.14
L16442.22
L16442.14EA
108
EUR
L16442.14
L16442.22
Cyclopentyltrimethoxysilane 95%
Cyclopentyltrimethoxysilane
Purity: 97%
CAS No.: 143487-47-2
Molecular Formula: C8H18O3Si
Formula Weight: 190.32
Flash Point: 54°(129°F)
Refractive Index: 1.4250
Boiling Point: 75°/10mm
Beilstein Reference: 8310738
UN No.: 1993
MDL No.: MFCD00236509
Formule:
C₈H₁₈O₃Si Kookpunt: 75 °C (10 mmHg) Dichtheid: 0,990 Vlampunt: 54 °C (129 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00236509 CAS nummer: 143487-47-2 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...