Colour coded inserts, CapInsert™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M957G M957Y M957L M957R M957B
SIMPM957GEA 24.4 EUR
SIMPM957G SIMPM957Y SIMPM957L SIMPM957R SIMPM957B
Colour coded inserts, CapInsert™
GLB-inzetstukken
These colour coded inserts could be applied to the tube closure, which eliminates jeopardising the integrity of your sample by transferring it to another vial.
  • Manually press fit and locks into place on top of closure