CapLock™ clips
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T330-75CLB T330-75CLG T330-75CLR T330-75CLW T330-75CLY
SIMPT330-75CLBEA 47.4 EUR
SIMPT330-75CLB SIMPT330-75CLG SIMPT330-75CLR SIMPT330-75CLW SIMPT330-75CLY
CapLock™ clips
Klemmen Klemmen voor algemeen gebruik
These CapLock™ clips are made of PE. They provide extra security against inadvertent cap openings on critical samples or during incubation above 80 °C.
  • Ideal for shipping samples
  • Suitable for 5 ml tubes with snap cap