Adapters for universal centrifuges
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T330AD15 T330AD50
SIMPT330AD15EA 37.3 EUR
SIMPT330AD15 SIMPT330AD50
Adapters for universal centrifuges
Centrifuge Tafelmodel centrifuges
These universal centrifuge adapters are made of acetal plastic.
  • Reusable and freezable (–90 °C)