Diantimoontrioxide analytical reagent, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Antimony trioxide, Diantimony trioxide, di-Antimony trioxide
M-Clarity™ quality level = MQ300
1.07836.0100 1.07836.1000
1.07836.0100EA 79.7 EUR
1.07836.0100 1.07836.1000
Diantimoontrioxide analytical reagent, Supelco®
Diantimoontrioxide
Formule: O₃Sb₂
MW: 291.52 g/mol
Kookpunt: 1425
Smeltpunt: 654
Dichtheid: 5.2 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011214
CAS nummer: 1309-64-4
EINECS: 215-175-0
UN: 1549
Merck Index: 13,00717
Order Now
Specification Test Results
Solubility 2.70 mg/l (20 °C)
Melting Point 656 °C (sublimed)
Molar Mass 291.49 g/mol
Bulk Density 800 - 1300 kg/m3
Boiling Point 1550 °C (1013 hPa)
Vapor Pressure 13.3 hPa (660 °C)
Density 5.4 g/cm3