Triphenyl phosphate 99+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
147670010 147672500 147670025 147670050
ACRO147670010EA 54.6 EUR
ACRO147670010 ACRO147672500 ACRO147670025 ACRO147670050
Triphenyl phosphate 99+%
Triphenyl phosphate
Formule: C₁₈H₁₅O₄P
Kookpunt: 370 °C (760 mmHg)
Smeltpunt: 47…53 °C
Dichtheid: 1,33 g/cm³ (20 °C)
Vlampunt: 220 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003031
CAS nummer: 115-86-6
EINECS: 204-112-2
UN: 3077
ADR: 9,III
Merck Index: 12,09872
Order Now
Specification Test Results
Appearance White Flakes and/or pastilles
Infrared spectrum Conforms
Melting point 47 to 53 °C
Water ≤0.1 %
Additional info Free phenols: 0.005%
GC ≥99.0 %
Color scale ≤50 APHA (after heat treatment)
Acid value ≤0.05 mg KOH/g