Triphenyl phosphate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Triphenylphosphate, TPP
L08130.30 L08130.0B
L08130.30EA 30.9 EUR
L08130.30 L08130.0B
Triphenyl phosphate ≥98%
Triphenyl phosphate
Purity: 98% CAS No.: 115-86-6 Molecular Formula: C18H15O4P Formula Weight: 326.29 Flash Point: 223°(433°F) Odour: Odourless Boiling Point: 247-250°/11mm Beilstein Reference: 1888236 Merck Reference: 14,9742 UN No.: 3077 MDL No.: MFCD00003031
Formule: C₁₈H₁₅O₄P
MW: 326.29 g/mol
Kookpunt: 247…250 °C (11 mmHg)
Smeltpunt: 48…52 °C
Dichtheid: 1.33 g/cm³ (20 °C)
Vlampunt: 223 °C (433 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003031
CAS nummer: 115-86-6
EINECS: 204-112-2
UN: 3077
ADR: 9 III
Merck Index: 12,09872