Tetramethylphosphonium bromide ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A17907.03 A17907.06
A17907.03EA 65.4 EUR
A17907.03 A17907.06
Tetramethylphosphonium bromide ≥98%
Tetramethylphosphonium bromide
Purity: 98% CAS No.: 4519-28-2 Molecular Formula: (CH3)4PBr Formula Weight: 171.03 MDL No.: MFCD00011802
Formule: C₄H₁₂BrP
MW: 171.02 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011802
CAS nummer: 4519-28-2