Carbachol 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
108240050
108240250
ACRO108240050EA
101
EUR
ACRO108240050
ACRO108240250
Carbachol 99%
Carbachol
Formule:
H₂NC(O)O(CH₂)₂N(CH₃)₃Cl Dichtheid: 1,389 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00012011 CAS nummer: 51-83-2 EINECS: 200-127-3 UN: 2811 ADR: 6.1,II Merck Index: 13,01786 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 200 to 204 °C (Dec) |
Loss on drying | ≤2.0 % (105 °C, 2 hrs) |
Titration with HClO4 | ≥98.5 % (On dried substance) |
UV | at 195 nm A: ≤1.0 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Sulfur compounds (as S) | ≤0.005 % |
Water soluble titr. acid | ≤0.0003 meq/g |
Thiophene | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...