Racks for tubes
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T450-25B T450-25O T450-25R
SIMPT450-25BEA 76.9 EUR
SIMPT450-25B SIMPT450-25O SIMPT450-25R
Racks for tubes
Rekken Tube Racks
Made of polypropylene.
  • Reusable, autoclavable and freezable (–90 °C)
  • Wells are numbered for easier identification
  • Good chemical resistance