Flash chromatography cartridges, CHROMABOND® Flash BT
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
732961 732962 732960 732963 732964 732965 732966 732967
MANA732961EA 202 EUR
MANA732961 MANA732962 MANA732960 MANA732963 MANA732964 MANA732965 MANA732966 MANA732967
Flash chromatography cartridges, CHROMABOND® Flash BT
Kolommen voor chromatografie
Ready-to-use Flash cartridges designed for use in the Biotage® Isolera™ systems without additional connectors or capillaries.
Safe hardware
- Heavy-duty polypropylene cartridges
- Pressure stability up to 15 bar (BT 4 - BT 120) and 12 bar (BT 200 - BT 330)
- Female Luer lock inlet / male Luer lock outlet
Robust phase
- Irregular unmodified silica SiOH (pore size 60 Å) with particle sizes of 40 – 63 µm