Tetrabutylammonium hydrogen sulphate ≥97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate
A14047.0B A14047.18 A14047.30
A14047.0BEA 490 EUR
A14047.0B A14047.18 A14047.30
Tetrabutylammonium hydrogen sulphate ≥97%
Tetrabutylammonium hydrogen sulphate
Purity: 97% CAS No.: 32503-27-8 Molecular Formula: [CH3(CH2)3]4NHSO4 Formula Weight: 339.54 Melting Point: 168-172° MDL No.: MFCD00011637
Formule: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
Smeltpunt: 168…172 °C
Dichtheid: 1.01 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011637
CAS nummer: 32503-27-8
EINECS: 251-068-5