Benzyltriethylammonium chloride for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Benzyl triethylammonium chloride
M-Clarity™ quality level = MQ200
8.10298.0100 8.10298.0500 8.10298.9040
8.10298.0100EA 38.7 EUR
8.10298.0100 8.10298.0500 8.10298.9040
Benzyltriethylammonium chloride for synthesis, Sigma-Aldrich®
Benzyltriethylammonium chloride
Formule: C₁₃H₂₂ClN
MW: 227.78 g/mol
Smeltpunt: 190…192 °C
Vlampunt: >100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011824
CAS nummer: 56-37-1
EINECS: 200-270-1
Order Now
Specification Test Results
Solubility 630 g/l (20 °C)
Melting Point 190 - 192 °C (decomposition)
Molar Mass 227.78 g/mol
Bulk Density 700 kg/m3
Flash Point 100 °C
pH 6 - 8 (100 g/l, H2O, 20 °C)
Ignition Point 300 °C