Tetraphenylphosphonium bromide ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A15860.09 A15860.18 A15860.22
A15860.09EA 37.4 EUR
A15860.09 A15860.18 A15860.22
Tetraphenylphosphonium bromide ≥98%
Tetraphenylphosphonium bromide
Purity: 98+% CAS No.: 2751-90-8 Molecular Formula: (C6H5)4PBr Formula Weight: 419.31 Melting Point: 296-302° Beilstein Reference: 3922383 Merck Reference: 14,9237 MDL No.: MFCD00011915
Formule: C₂₄H₂₀BrP
MW: 419.3 g/mol
Smeltpunt: 296…302 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011915
CAS nummer: 2751-90-8
EINECS: 220-393-4
Order Now
Specificaties
Environmentally Preferable