Ammoniumchloride ≥99.5%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Ammonium muriate, Salmiac
Purity: 99.5% min
CAS No.: 12125-02-9
Molecular Formula: NH4Cl
Formula Weight: 53.49
Physical Form: Granular
Density: ≈1.53
Refractive Index: 1.642
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 340° subl.
Solubility: Soluble in water, methanol, ethanol. Insoluble in acetone, ether, ethyl acetate.
Merck Reference: 14,509
MDL No.: MFCD00011420
As a flux in galvanizing steel sheet, in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton
Formule:
NH₄Cl MW: 53.49 g/mol Smeltpunt: 340 °C (sublimatie) Dichtheid: 1.53 g/cm³ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011420 CAS nummer: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |