Ammoniumchloride 98
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ammonium muriate, Salmiac
A15000.0B A15000.0I A15000.30 A15000.36
A15000.0BEA 49.5 EUR
A15000.0B A15000.0I A15000.30 A15000.36
Ammoniumchloride 98
Ammoniumchloride
Purity: 98+% CAS No.: 12125-02-9 Molecular Formula: H4ClN Formula Weight: 53.49 Refractive Index: 1.642 Odour: Odourless Beilstein Reference: 4371014 Merck Reference: 14,509 MDL No.: MFCD00011420
Formule: NH₄Cl
MW: 53.49 g/mol
Smeltpunt: 340 °C (sublimatie)
Dichtheid: 1.53
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011420
CAS nummer: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM