Ammonium iodide ≥99.995% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
40255.06
40255.14
40255.22
40255.06EA
85.8
EUR
40255.06
40255.14
40255.22
Ammonium iodide ≥99.995% (metals basis)
Ammonium iodide
Purity: 99.995%
CAS No.: 12027-06-4
Molecular Formula: H4IN
Formula Weight: 144.94
Physical Form: -4 Mesh
Density: 2.514
Refractive Index: 1.7031
Sensitivity: Light and Air Sensitive and Hygroscopic
Appearance: White to pale yellow
Odour: Odourless
Melting Point: 551° subl.
Merck Reference: 14,529
MDL No.: MFCD00011424
Formule:
NH₄I Dichtheid: 2,514 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011424 CAS nummer: 12027-06-4 EINECS: 234-717-7 Merck Index: 14,529 |