Ammonium iodate ≥99%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 99%
CAS No.: 13446-09-8
Molecular Formula: H4INO3
Formula Weight: 192.94
Physical Form: Crystalline
Density: 3.309
Appearance: White
Odour: Odourless
Melting Point: 150°
UN No.: 1479
MDL No.: MFCD00049684
Formule:
H₄INO₃ Smeltpunt: 150 °C Dichtheid: 3,309 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00049684 CAS nummer: 13446-09-8 EINECS: 236-592-4 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...