Diammoniumperoxodisulfaat (APS)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

9178.1 9178.2
ROTH9178.1EA 0 EUR
ROTH9178.1 ROTH9178.2
Diammoniumperoxodisulfaat (APS)
Diammoniumperoxodisulfaat (APS)
Formule: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Smeltpunt: 120
Dichtheid: 1.98 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003390
CAS nummer: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1 III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR