Diammoniumperoxodisulfaat (APS)
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Peroxidisulphuric acid diammonium salt, Ammonium persulphate
9592.1
9592.2
9592.3
ROTH9592.1EA
0
EUR
ROTH9592.1
ROTH9592.2
ROTH9592.3
Diammoniumperoxodisulfaat (APS)
Diammoniumperoxodisulfaat (APS)
Formule:
(NH₄)₂S₂O₈ Smeltpunt: 120 °C Dichtheid: 1,98 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003390 CAS nummer: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 13,00545 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...