Dimethylsulfoxide ≥99.9% for HPLC
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
Synoniemen:
Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
22914.K2
22914.0F
22914.M1
22914.K2EA
179
EUR
22914.K2
22914.0F
22914.M1
Dimethylsulfoxide ≥99.9% for HPLC
Dimethylsulfoxide
Purity: 99.9+%
CAS No.: 67-68-5
Molecular Formula: (CH3)2SO
Formula Weight: 78.13
Flash Point: 87°(189°F)
Physical Form: Liquid
Density: 1.101
Refractive Index: 1.4790
Sensitivity: Hygroscopic
Odour: Mild
Melting Point: 18.4°
Boiling Point: 189°
Beilstein Reference: 506008
Merck Reference: 14,3259
MDL No.: MFCD00002089
Formule:
(CH₃)₂SO MW: 78.14 g/mol Kookpunt: 189 °C Smeltpunt: 18.4 °C Dichtheid: 1.101 Vlampunt: 87 °C (189 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002089 CAS nummer: 67-68-5 EINECS: 200-664-3 Merck Index: 14,3259 |