Dimethylsulfoxide ≥99.9% ACS
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
Purity: 99.9% min
CAS No.: 67-68-5
Molecular Formula: C2H6OS
Formula Weight: 78.13
Flash Point: 87°(189°F)
Physical Form: Clear liquid
Density: 1.101
Refractive Index: 1.4790
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Mild
Melting Point: 18.4°
Boiling Point: 189°
Beilstein Reference: 506008
Merck Reference: 14,3259
MDL No.: MFCD00002089
Formule:
(CH₃)₂SO Kookpunt: 189 °C Dichtheid: 1,101 Vlampunt: 87 °C (189 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002089 CAS nummer: 67-68-5 EINECS: 200-664-3 Merck Index: 14,3259 |