Dimethylsulfoxide, EMPLURA®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
MQ200
1.16743.1000 1.16743.6025
1.16743.1000EA 69 EUR
1.16743.1000 1.16743.6025
Dimethylsulfoxide, EMPLURA®, Supelco®
Dimethylsulfoxide
Formule: (CH₃)₂SO
MW: 78.14 g/mol
Kookpunt: 189
Smeltpunt: 18.5
Dichtheid: 1.101 g/cm³ (20 °C)
Vlampunt: 87
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002089
CAS nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM