Dimethylsulfoxide
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
4720.1
4720.2
4720.3
4720.4
ROTH4720.1EA
0
EUR
ROTH4720.1
ROTH4720.2
ROTH4720.3
ROTH4720.4
Dimethylsulfoxide
Dimethylsulfoxide
Formule:
(CH₃)₂SO Kookpunt: 189 °C (1013 hPa) Dichtheid: 1,101 g/cm³ (20 °C) Vlampunt: 87 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002089 CAS nummer: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...