Volumetric flasks, Class A, glass, clear, beaded rim, with DAkkS certificate, BLAUBRAND®
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DAKKS37049 DAKKS37050
BRNDDAKKS37049EA 83 EUR
BRNDDAKKS37049 BRNDDAKKS37050
Volumetric flasks, Class A, glass, clear, beaded rim, with DAkkS certificate, BLAUBRAND®
Kolven Maatkolven
Borosilicate glass 3.3, with beaded rim.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
Certificaten: DIN EN ISO 1042
Bestelinformatie: Supplied with a DAkkS certificate.