Dioctyl sulphide ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Dioctylsulphane

O0108-10G O0108-100G
TCIAO0108-10GEA 17.6 EUR
TCIAO0108-10G TCIAO0108-100G
Dioctyl sulphide ≥97.0% (by GC)
Dioctyl sulphide
Formule: C₁₆H₃₄S
MW: 258.51 g/mol
Kookpunt: 309°C
Vlampunt: 160°C
MDL Nummer: MFCD00009566
CAS nummer: 2690-08-6
EINECS: 220-259-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c63ea24b35cf3af841e71bbfa2cc5d4bfbc4f97c&vid=c63ea24b35cf3af841e71bbfa2cc5d4bfbc4f97c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR