Diphenyl sulphide 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
130950250 130951000
ACRO130950250EA 39.2 EUR
ACRO130950250 ACRO130951000
Diphenyl sulphide 99%
Diphenyl sulphide
Formule: C₁₂H₁₀S
Kookpunt: 296 °C (1013 hPa)
Smeltpunt: –40 °C
Dichtheid: 1,11 g/cm³
Vlampunt: >112 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003064
CAS nummer: 139-66-2
EINECS: 205-371-4
ADR: 6.1,III
Merck Index: 12,07467
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6317 to 1.6337 (20°C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.001 %