Iodine monochloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
196961000 196965000
ACRO196961000EA 67 EUR
ACRO196961000 ACRO196965000
Iodine monochloride 98%
Iodine monochloride
Formule: ClI
Kookpunt: 97 °C (760 mmHg)
Smeltpunt: 26 °C
Dichtheid: 3,2 g/cm³
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011354
CAS nummer: 7790-99-0
EINECS: 232-236-7
UN: 1792
ADR: 8,II
Merck Index: 13,05039
Order Now
Specification Test Results
Appearance Dark red-brown to brown to black Liquid or solid
Titration Iodometric ≥97.5 %