Fenylhydrazine ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A11246.0B
A11246.18
A11246.30
A11246.0BEA
143
EUR
A11246.0B
A11246.18
A11246.30
Fenylhydrazine ≥97%
Fenylhydrazine
Purity: 97%
CAS No.: 100-63-0
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 88°(190°F)
Physical Form: Liquid
Density: 1.099
Refractive Index: 1.6065
Sensitivity: Air and Light Sensitive
Appearance: Yellow
Melting Point: 18-20°
Boiling Point: 238-241°
Solubility: Soluble in dilute acids
Beilstein Reference: 606080
Merck Reference: 14,7293
UN No.: 2572
MDL No.: MFCD00007573
Formule:
C₆H₈N₂ MW: 108.14 g/mol |
MDL Nummer:
MFCD00007573 CAS nummer: 100-63-0 Merck Index: 12,07447 |