Manual crimping tools, advanced ergonomic version
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
735211 735220 735311 735320 735208
MANA735211EA 273 EUR
MANA735211 MANA735220 MANA735311 MANA735320 MANA735208
Manual crimping tools, advanced ergonomic version
Capsuleer- en decapsuleertangen
These manual crimping tools with ergonomically designed handles are more lightweighted than complete steel crimpers.
Ergonomic crimpers:
• Adjustable knob on the crimping head that is easily accessible and visible
• Activated by bottom handle motion only which allows a steadier and safer hold of the tool during crimping
• Crimping head design and alignment makes better vertical clearance over the vial
Ergonomic decappers:
• Allow safe removal of caps
• No adjustment required