3-Methylindole 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
127030100 127030250 127031000
ACRO127030100EA 60.6 EUR
ACRO127030100 ACRO127030250 ACRO127031000
3-Methylindole 98%
3-Methylindole
Formule: C₉H₉N
Kookpunt: 265…266 °C (755 mmHg)
Smeltpunt: 95…98 °C
Dichtheid: 1,003 g/cm³ (99.6 °C)
Vlampunt: 132 °C
MDL Nummer: MFCD00005627
CAS nummer: 83-34-1
EINECS: 201-471-7
UN: 3077
Merck Index: 12,08700
Order Now
Specification Test Results
Appearance White to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 95 to 98 °C
GC ≥97.5 %
Color scale ≤50 APHA
Impurity ≤1.1 % Tosyl chloride