Europium(III) oxide ≥99.9%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36352.04 36352.09
36352.04EA 198 EUR
36352.04 36352.09
Europium(III) oxide ≥99.9%
Europium(III) oxide
Purity: 99.9% CAS No.: 1308-96-9 Molecular Formula: Eu2O3 Formula Weight: 351.92 Physical Form: 3-12mm Sintered Lump Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00010997
For vacuum deposition
Formule: Eu₂O₃
MW: 351.92 g/mol
Smeltpunt: 2291 °C
Dichtheid: 7.42 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010997
CAS nummer: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,3904