Holmium(III) oxide ≥99.9%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Diholmium trioxide, Holmium oxide
36357.09 36357.18
36357.09EA 197 EUR
36357.09 36357.18
Holmium(III) oxide ≥99.9%
Holmium(III) oxide
Purity: 99.9% CAS No.: 12055-62-8 Molecular Formula: Ho2O3 Formula Weight: 377.86 Physical Form: 3-12mm Sintered Lump Odour: Odourless Merck Reference: 14,4727 MDL No.: MFCD00011053
Formule: Ho₂O₃
MW: 377.86 g/mol
Kookpunt: 2600
Smeltpunt: 2360 °C
Dichtheid: 3.79 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011053
CAS nummer: 12055-62-8
EINECS: 235-015-3