Safety shoes, BOA® Fit System, jo_Max BOA® Low, Jori
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12771/38 12771/39 12771/40 12771/41 12771/42 12771/43 12771/44 12771/45 12771/46 12771/47 12771/48 12771/36 12771/37
ELTN12771/38EA 85.9 EUR
ELTN12771/38 ELTN12771/39 ELTN12771/40 ELTN12771/41 ELTN12771/42 ELTN12771/43 ELTN12771/44 ELTN12771/45 ELTN12771/46 ELTN12771/47 ELTN12771/48 ELTN12771/36 ELTN12771/37
Safety shoes, BOA® Fit System, jo_Max BOA® Low, Jori
Schoenen en laarzen
Antistatic safety shoe made from hydrophobised nubuck leather, mesh inserts and reflective materials, skin-friendly textile lining, a full, padded bellows tongue and collar, and a dual-density rubber/PU jo_MAX sole (3,0 mm profile, oil, fuel and chemical resistant, temperature resistant from approx. −20 and +200 °C).
  • Climate regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full-length inlay sole
  • Lace- and buckle-free Boa® closure system - secure, quick and easy by turning nob
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaten: EN ISO 20345; S3, SRC